Learn More
Cayman Chemical FlavokawaIn B 10mg
SDP
Supplier:  Cayman Chemical 1965210 MG
Encompass_Preferred
A natural chalcone that induces apoptosis in prostate cancer cell lines (IC50s = 32, 48, 6.2, and 3.9 µM for LAPC4, LNCaP, PC-3, and DU145 cells, respectively, treated for 48 hours); increases Bim expression and inhibits growth of DU145 xenografts in mice; hepatotoxic
Catalog No. 50-298-1387