Learn More
Medchemexpress LLC HY-N2132 10mg Medchemexpress, Flavokawain B CAS:1775-97-9 Purity:>98%
Supplier: Medchemexpress LLC HYN213210MG
Medchemexpress, HY-N2132 10mg Flavokawain B CAS:1775-97-9 Flavokawain B (Flavokavain B) is a chalcone isolated from the root extracts of kava-kava plant and a potent apoptosis inducer for inhibiting the growth of various cancer cell lines. Flavokawain B (Flavokavain B) shows strong antiangiogenic activity. Flavokawain B (Flavokavain B) inhibits human brain endothelial cell (HUVEC) migration and tube formation with very low and non-toxic concentrations. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.