Learn More
Medchemexpress LLC Flavopiridol | 146426-40-6 | >=95.0% | 401.84 | C21H20ClNO5 | 1 G
SDP
Supplier:  Medchemexpress LLC HY100051G
Encompass_Preferred
Flavopiridol (Alvocidib) is a small-molecule, ATP-competitive cyclin-dependent kinase (CDK) inhibitor used in research to study cell cycle regulation, transcriptional control, and apoptosis. It exhibits potent inhibition of multiple CDKs and is supplied for laboratory and biochemical assays.
  • Broad-spectrum, ATP-competitive inhibition of cyclin-dependent kinases for cell cycle studies.
  • Potent activity with reported IC50 values of 30 nM (CDK1), 170 nM (CDK2), and 100 nM (CDK4).
  • Applicable to apoptosis and transcription regulation research involving CDK pathways.
  • Provided as a characterized research chemical with documented CAS number and molecular weight.
  • Suitable for in vitro biochemical assays and cellular studies when handled per safety guidance.
Catalog No. 50-002-15369