Learn More
Enzo Life Sciences Flavopiridol (25mg). CAS: 146426-40-6
SDP
Supplier:  Enzo Life Sciences ALX430161M025
Encompass_Preferred
Potent inhibitor of cyclin-dependent kinases (CDKs). Induces apoptosis in certain tumor cells. Interacts with multidrug resistance protein 1 (MRP1). Alternative name: Alvocidib, (-)-2-(2-Chlorophenyl)-5,7-dihydroxy-8-[(3R,4S)-3-hydroxy-1-methyl-4-piperidinyl]-4H-1-benzopyran-4-one. Purity: ≥97% (HPLC). Appearance: Yellow powder. Solubility: Soluble in chloroform; slightly soluble in water. Long Term Storage: +4°C.
Catalog No. 50-200-9251