Learn More
Selleck Chemical LLC Flavopiridol (Alvocidib) S1230-10mM/1mL
SDP
Supplier:  Selleck Chemical LLC S123010MM/1ML
Encompass_Preferred
Flavopiridol (Alvocidib) competes with ATP to inhibit CDKs including CDK1 CDK2 CDK4 CDK6 and CDK9 with IC50 values in the 20-100 nM range It is more selective for CDK1 2 4 6 9 versus CDK7 Flavopiridol is initially found to inhibit EGFR and PKA Flavopiridol induces autophagy and ER stress Flavopiridol blocks HIV-1 replication Phase 1/2
Catalog No. 50-311-1210