Learn More
Medchemexpress LLC Flavopiridol hydrochloride | 131740-09-5 | MFCD26792554 | 438.30 g/mol | C21H21Cl2NO5 | 500 MG
SDP
Supplier:  Medchemexpress LLC HY10006500MG
Encompass_Preferred
Flavopiridol hydrochloride (alvocidib hydrochloride) is a broad-spectrum ATP-competitive inhibitor of cyclin-dependent kinases used in biochemical and cell-based research to study cell cycle and transcription. Reported IC50 values include CDK1 = 30 nM, CDK2 = 170 nM, and CDK4 = 100 nM. The compound is soluble in DMSO and water and has documented storage recommendations.
  • Broad-spectrum CDK inhibitor with low-nanomolar potency.
  • Useful for cell cycle and transcription studies in vitro.
  • Soluble in DMSO and water at research concentrations.
  • Documented solubility in common solvents and mixed vehicles for formulation.
  • Stable when stored sealed; solution stability noted at -80°C (1 year) and -20°C (6 months).
Catalog No. 50-002-15430