Learn More
Medchemexpress LLC Flavopiridol hydrochloride (alvocidib hydrochloride) | 131740-09-5 | 99.7% | 438.30 | C21H21Cl2NO5 | 50 MG
SDP
Supplier:  Medchemexpress LLC HY1000650MG
Encompass_Preferred
Flavopiridol hydrochloride (alvocidib hydrochloride) is a research-grade, ATP-competitive inhibitor of cyclin-dependent kinases used in cell-cycle and signal-transduction studies. Supplied as a hydrochloride salt, it is formulated for biochemical and cellular assays and offers documented solubility and storage recommendations for reliable experimental use.
  • Inhibits CDK1, CDK2, and CDK4 with reported IC50s of 30 nM, 170 nM, and 100 nM.
  • High purity confirmed by HPLC suitable for research applications.
  • Soluble in DMSO and water at ≥ 20 mg/mL for convenient stock preparation.
  • Provided as a solid with recommended sealed, low-temperature storage to maintain stability.
  • Available with solvent option to simplify assay setup.
Catalog No. 50-002-15653