Learn More
Medchemexpress LLC Flavopiridol hydrochloride (alvocidib hydrochloride) | 131740-09-5 | MFCD26792554 | 99.7% | 438.30 | C21H21Cl2NO5 | 1 ML
SDP
Supplier:  Medchemexpress LLC HY1000610MM 1ML
Encompass_Preferred
Flavopiridol hydrochloride (alvocidib hydrochloride) is an ATP-competitive inhibitor of cyclin-dependent kinases used in research to probe cell cycle regulation, autophagy, and HIV-related pathways. It inhibits CDK1, CDK2, and CDK4 with reported IC50 values of approximately 30 nM, 170 nM, and 100 nM, respectively. Supplied as a solid for solution preparation, it has a molecular weight of 438.30 and high purity for laboratory experiments.
  • ATP-competitive inhibitor of cyclin-dependent kinases
  • Inhibits CDK1, CDK2, and CDK4 with reported IC50s of ~30, 170, 100 nM
  • High purity suitable for research applications
  • Supplied as solid for preparation of working solutions
  • Useful for studies of cell cycle, autophagy, and HIV-related pathways
Catalog No. 50-002-15490