Learn More
Sorbent Technologies Inc Flash Cartridges, Spherical Silica Gel, Premium Rf, 70A, 20-45um, 80g, 10/pk
SDP
Supplier:  Sorbent Technologies Inc CFC7260008010
Encompass
Purity™ Flash Cartridges are superior because they're packed with Premium Rf™ Spherical Silica Gel. The porosity is 70A, particle distribution is 20-45um, bed mass is 80g, and quantity is 10 per pack. Sorbtech's packing technique and Premium Rf's narrow particle distribution results in a uniform packed bed. The benefits are no channeling, higher resolution, decrease in backpressure, faster flow, and faster run time. Additionally, the small pore volume allows for more silica gel per column volume leading to greater loadability and less solvent use. Finally, the high purity silica gel (minimal surface metals) reduces mixed-mode interactions (no peak tailing or resolution loss). The polypropylene cartridge has standard luer-lock end fittings and is spin-welded (leak-free) allowing higher pressures up to 350 psi. The cartridges are compatible with all flash chromatography systems.
Catalog No. 50-113-9143