Learn More
Cole-Parmer DWK Life Sciences (Kimble) 14035-1200 BEAKERplus Combination Beaker and Flask, 1200 mL, 2/pk
Supplier:  Cole-Parmer 3451708
Encompass_Preferred
Kimax BeakerPlus™ Combination Beaker and Flask is ideal for mixing and for use with funnels or filters. The unique design features a wide fluted rim with pour spout and narrow recessed neck to reduce splashing during agitation. Manufactured with borosilicate glass, these beaker/flasks offer excellent chemical and heat resistance.
Catalog No. NC1911929