Learn More
Medchemexpress LLC Flecainide-d4 (acetate) | 1276197-21-7 | 99.0% | 478.42 | 1 MG
SDP
Supplier:  Medchemexpress LLC HY17429S1MG
Encompass_Preferred
Flecainide-d4 (acetate) is the deuterium labeled Flecainide acetate, a class 1C antiarrhythmic agent used for managing supraventricular arrhythmia. It functions by blocking the Nav1.5 sodium channel in the heart, leading to a prolongation of the cardiac action potential.
  • Used as a tracer
  • Used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS
Catalog No. 50-003-86418