Learn More
Medchemexpress LLC Flecainide hydrochloride | 57415-44-8 | 99.9% | 450.80 | 50 MG
SDP
Supplier:  Medchemexpress LLC HYW010950A50MG
Encompass_Preferred
Flecainide hydrochloride is an orally active antiarrhythmic agent. It works by blocking sodium channels and inhibiting calcium ion release mediated by the cardiac ryanodine receptor (RyR2). This product is used in the research of diseases such as catecholaminergic polymorphic ventricular tachycardia (CPVT).
  • Orally active antiarrhythmic agent
  • Blocks sodium channels
  • Inhibits calcium ion release mediated by cardiac ryanodine receptor (RyR2)
  • Used in research for conditions like catecholaminergic polymorphic ventricular tachycardia (CPVT)
Catalog No. 50-004-52891