Learn More
Ace Glass, Inc. Chuck, Flex-Grip;, 6mm stir shaft to 1/4in motor
SDP
Supplier:  Ace Glass, Inc. 812404
Encompass_Preferred
Chuck, Flex-Grip;, 6mm stir shaft to 1/4in motor
Catalog No. NC9267020