Learn More
MSC Flexbar Surface Roughness Standards Set, 30 Specimens, Nickel, 2 to 500 AA micro Inch
Supplier:  MSC 06558142
Encompass
This surface roughness standards set provides 30 electroformed solid nickel specimens for visually and tactilely comparing surface finishes. The set covers a range from 2 to 500 AA micro inches and includes various machining processes, conforming to SAE and military specifications.
  • 30 specimens, each 7/8 in x 3/8 in.
  • Made of electroformed solid nickel.
  • Accurately reproduces various surface finishes for visual and tactile comparison.
  • Includes instructions and a case.
  • Enables quick comparisons, saving time and preventing scrap.
  • Markings in AA (microinches) and Metric Ra (micrometers).
  • Conforms to SAE and military specifications.
  • Surface finishes from 2 to 500 AA micro "
  • Covers reaming, milling, turning, grinding, and flat lapping.
Catalog No. 50-005-28448