Learn More
Sartorius Flexboy 2D Bag, 10 L, Sartopore 2 0.45 | 0.2 m connector inlet, "" TC sanitary flange connector outlet, "" ID Clear C-Flex 500 mm line outlet, Clave connector sampling, 20 pcs.
Supplier:  Sartorius FFB110911/DEL
Encompass
Flexboy Bioprocessing Bags are designed for the preparation, storage and transport of biopharmaceutical solutions, intermediates and final bulk products. They provide a single-use alternative to traditional glass, stainless steel and rigid plastic carboys in a large variety of applications. Flexboy Bags are available in bag chamber volumes between 5 ml and 50 l. They are supplied sterilised and ready to use. Multiple configurations that also integrate thermoweldable TPE tubing are provided for flexible incorporation into your process.
Catalog No. NC0311294