Learn More
Braintree Scientific Flexible Sterile Disposable Feeding Needle (100/bx)
SDP
Supplier:  Braintree Scientific DT9928
Encompass
Flexible PTFE Disposable Feeding Needle. Plastic (PTFE) animal feeding needles (AFN) allow user to precisely dispense liquid to animal while minimizing the chances of esophageal piercing. sterile, non-toxic, and non-pyrogenic, available in a variety of gauges and lengths. **Sold for research purposes only and not for human use**
Catalog No. 50-195-4670