Learn More
Apexbio Technology LLC FLI-06 313967-18-9 25mg
SDP
Supplier:  Apexbio Technology LLC B308325
Encompass_Preferred
FLI-06 (CAS 313967-18-9) is a small molecule inhibitor of the Notch signaling pathway exhibiting an EC50 of 2 3 M Notch signaling regulates cell fate decisions and its dysregulation is implicated in oncogenesis and developmental disorders FLI-06 impedes Notch E-eGFP accumulation and reduces NICD-eGFP production in HeLa cells with effects that are fully reversible upon washout indicating low cytotoxicity Additionally FLI-06 significantly decreases A secretion upstream of - and -secretase cleavage events and disrupts Golgi structure by interfering with early secretory pathway membrane trafficking This compound is valuable for studying Notch signaling secretory pathway dynamics and related cellular processes
Catalog No. 50-000-94148