Learn More
Medchemexpress LLC Flibanserin-d4-1 | 2122830-91-3 | 99.3% | C20H17D4F3N4O | 1 MG
SDP
Supplier:  Medchemexpress LLC HYA0095S11MG
Encompass_Preferred
Flibanserin-d4-1 is a deuterium-labeled version of Flibanserin. Flibanserin (BIMT-17) is a full agonist of the serotonin 5-HT1A receptor (Ki=1 nM) and an antagonist of 5-HT2A (49 nM). It binds to dopamine D4 receptors (4-24 nM) and has negligible affinity for other neurotransmitter receptors and ion channels. Flibanserin is effective in treating hypoactive sexual desire disorder (HSDD).
  • Suitable for research applications.
  • Can be used as a tracer.
  • Internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
  • Incorporates stable heavy isotopes for quantitation during drug development.
  • Deuteration can impact pharmacokinetic and metabolic profiles of drugs.
Catalog No. 50-004-32196