Learn More
Aviva Systems Biology Corp FLJ33706 Antibody - C-terminal region (ARP36270_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP36270_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against FLJ33706. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the C terminal region of human FLJ33706. Alternate names: ApplicationWB. Species reactivity: Human
Catalog No. 50-014-56098