Learn More
Aviva Systems Biology Corp FLJ35767 Antibody - middle region (ARP56005_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP56005_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against FLJ35767. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human FLJ35767. Alternate names: ApplicationWB. Species reactivity: Cow, Human, Mouse, Rat
Catalog No. 50-014-61692