Learn More
MSC Cortech USA Polyethylene Basket Truck, Flame Retardant, 15 Bushel, 1000 lb
Supplier:  MSC 38196754
Encompass
The Kangaroo Cart is a durable, rotationally molded polyethylene basket truck designed for various material handling applications. This model is flame retardant, with a 15-bushel capacity and a 1,000 lb load rating, making it suitable for intense use in environments like laundry, waste collection, and general material transport.
  • Constructed from high-impact, no-break polyethylene for enhanced durability
  • Features a flame-retardant design, passing Cal 133 standards
  • Offers a 15-bushel capacity and a 1,000 lb load rating
  • Equipped with heavy-duty casters secured with stainless steel, tamper-proof hardware
  • Includes push-pull handles on both sides to accommodate utility use
  • Designed with a drain plug for easy cleaning and maintenance
  • Built with a .200 nominal wall thickness and span design for increased rigidity
Catalog No. 50-283-8188