Learn More
Aviva Systems Biology Corp FLNC Antibody - N-terminal region (ARP64454_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP64454_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against FLNC. It was validated on Western Blot. The immunogen is a synthetic peptide directed towards the N-terminal region of Human FLNC. Alternate names: ABP-280, ABP280A, ABPA, ABPL, FLJ10186, FLN2ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Zebrafish
Catalog No. 50-288-265