Learn More
IDEXX LPD FLOCK CHEK CHICKEN ANEMIA KIT, ELISA, Blocking ELISA, Semi-quantitative, 5 plates, 480 samples, 2-7 °C, Worldwide availability
Supplier: IDEXX LPD 9908702
The IDEXX CAV Ab Test is an enzyme-linked immunosorbent assay (ELISA) intended for the detection of antibodies to Chicken Anemia Virus (CAV) in chicken serum, facilitating flock screening and vaccination monitoring. This testing kit streamlines the process of health assessments in poultry, enabling better management and production decisions.
- Assay principle is ELISA.
- Targets antibodies against Chicken Anemia Virus.
- Sample type is chicken serum.
- Test format is blocking ELISA.
- Produces semi-quantitative results.
- Detection dilution is 1:10.
- Vaccine response dilution for titer calculation is 1:100.
- Kit size includes 5 plates/strips.
- Maximum samples that can be processed is 480, with 5 x 96-well plates.
- Recommended storage temperature is 2-7 °C.
- Available worldwide.
- Produced by IDEXX LPD.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.