Learn More
Medchemexpress LLC Flonicamid | 158062-67-0 | 98.0% | 100 MG

Supplier: Medchemexpress LLC HY119649100MG
Flonicamid is an analytical standard of the insecticide flonicamid supplied as a 100 MG solid for research and analytical applications. It has molecular formula C9H6F3N3O, molecular weight 229.16 g/mol, CAS 158062-67-0, and a stated purity of 98.0%. The material is intended for analytical and laboratory use and should be stored according to the certificate of analysis.
- Analytical standard for pesticide residue and insecticide research.
- Stated purity 98.0% for reliable analytical performance.
- Solid form; store under conditions specified in the certificate of analysis.
- Molecular weight 229.16 g/mol; formula C9H6F3N3O.
- Packaged as a 100 MG unit for laboratory use.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.