Learn More
Medchemexpress LLC Flonicamid | 158062-67-0 | 99.9% | 250 MG

Supplier: Medchemexpress LLC HY119649250MG
Flonicamid is a pyridinecarboxamide-class systemic insecticide provided as an analytical standard for research and formulation work. It is supplied as an off-white to light beige solid with high reported purity and is suitable for analytical testing and laboratory studies.
- Systemic insecticide active against piercing-sucking pests.
- High purity analytical standard (reported purity 99.87%).
- Molecular formula C9H6F3N3O; molecular weight 229.16 g/mol.
- Supplied in small-pack quantities for laboratory use.
- Soluble in dimethyl sulfoxide (DMSO).
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.