Learn More
Medchemexpress LLC Flonoltinib TFA | 2928093-29-0 | 98.0% | 250 MG
SDP
Supplier:  Medchemexpress LLC 250MGHY130247A
Encompass_Preferred
Flonoltinib TFA is a potent and orally active dual JAK2/FLT3 inhibitor with IC50 values of 0.7 nM, 4 nM, 26 nM and 39 nM for JAK2, FLT3, JAK1 and JAK3, respectively. It exhibits anti-cancer activity.
  • Potent and orally active dual JAK2/FLT3 inhibitor
  • Exhibits anti-cancer activity
  • Inhibits JAK2 (0.7 nM), FLT3 (4 nM), JAK1 (26 nM), JAK3 (39 nM)
  • Down-regulates p-FLT3 in a dose-dependent manner
  • Induces apoptosis in MV4-11 cells
  • Induces cell cycle arrest with a G1/G0 percentage of 85% at 100 nM in MV4-11 cells
  • Exhibits significant antitumor effects in mouse models
Catalog No. 50-003-42087