Learn More
Chemglass Life Sciences Floor Dolly for Huber Ministat, TFE Coated
SDP
Supplier:  Chemglass Life Sciences CG1978M600
Encompass
Floor Dolly for Huber Ministat, TFE Coated. Floor model dolly is designed for use with Huber CG-1978-M Ministat or other brands of circulators. The dolly has 3 diameter wheels, the two in the front being swivel style. Slotted tray is coated with Green TFE and an optional spill tray which fits into the tray is available. An optional 36 long pull handle is also available.
Catalog No. 50-153-1357