Learn More
Medchemexpress LLC Florasulam | 145701-23-1 | 99.4% | 10 G
SDP
Supplier:  Medchemexpress LLC HYB184510G
Encompass_Preferred
Florasulam is a targeted post-emergent herbicide that belongs to the triazolopyrimidine sulfonanilide class. It functions by inhibiting acetolactate synthase (ALS) in plants, disrupting plant cell division, reducing growth, and ultimately leading to plant death. ALS is located in the chloroplasts and plays a crucial role in the biosynthesis of branched-chain amino acids.
  • Targeted post-emergent herbicide
  • Inhibits acetolactate synthase (ALS) in plants
  • Disrupts plant cell division and reduces growth
Catalog No. 50-004-09778