Learn More
Medchemexpress LLC Florbetapir styrylpyridine tosylate precursor | 1205550-99-7 | 99.7% | 1 ML
SDP
Supplier:  Medchemexpress LLC HY12094710MM 1ML
Encompass_Preferred
This compound is a tosylate precursor used to synthesize 18F-labeled florbetapir derivatives for positron emission tomography (PET) imaging of neurodegenerative disease. Supplied as a 10 mM solution in DMSO, it facilitates radiofluorination through a primary tosylate leaving group that is readily removed during chromatographic purification.
  • Precursor for 18F radiofluorination to produce PET imaging agents.
  • Supplied as a 10 mM solution in DMSO for direct use in radiochemistry.
  • High chemical purity suitable for radiolabeling workflows.
  • Primary tosylate leaving group enables efficient radiofluorination and removal.
  • Stable when stored frozen; follow supplier storage recommendations.
Catalog No. 50-002-32774