Learn More
Rockland Immunochemicals Anti-MOUSE IgG (H&L) (SHEEP) Antibody Fluorescein Conjugated - 610-602-002
SDP
Supplier:  Rockland Immunochemicals 610602002
Encompass
Anti-Mouse IgG Fluorescein Antibody generated in sheep detects reactivity to Mouse IgG. When choosing a secondary antibody product, consideration must be given to species and immunoglobulin specificity, conjugate type, fragment and chain specificity, level of cross-reactivity, and host-species source and fragment composition. This product is designed for immunofluorescence microscopy, fluorescence based plate assays (FLISA) and fluorescent western blotting. This product is also suitable for multiplex analysis, including multicolor imaging, utilizing various commercial platforms.
Catalog No. NC9912239