Learn More
Medchemexpress LLC Florfenicol | 73231-34-2 | 99.1% | 250 MG
SDP
Supplier:  Medchemexpress LLC 250MGHYB1374R
Encompass_Preferred
Florfenicol is an orally active broad-spectrum antibacterial antibiotic with anti-inflammatory, proapoptotic, and immunomodulatory functions.
  • Inhibits fibroblast proliferation and autophagy by inducing mitochondrial dysfunction.
  • Exhibits cytotoxic effects on *Unio crusus* and *Cyprinus carpio* primary cell lines.
  • Shows toxicity to developing chicks and can induce early embryonic death.
  • Induces renal inflammatory response and cell apoptosis in chicks.
  • Exhibits regulatory effects on lymphocyte subsets and humoral immune responses.
  • Inhibits allergic airway inflammation in mice.
  • Can inhibit humoral and cellular immune responses in mice.
Catalog No. 50-004-14411