Learn More
Medchemexpress LLC Florfenicol (standard) | 73231-34-2 | 99.72% | 50 MG
SDP
Supplier:  Medchemexpress LLC HYB1374R50MG
Encompass_Preferred
Florfenicol (Standard) is the analytical standard of Florfenicol. It is an orally active broad-spectrum antibacterial antibiotic with anti-inflammatory, pro apoptotic, and immunomodulatory functions. This analytical standard is commonly used in qualitative, quantitative and methodological research experiments in HPLC, GC and MS.
  • Intended for research and analytical applications
  • Broad-spectrum antibacterial antibiotic
  • Anti-inflammatory, pro apoptotic, and immunomodulatory functions
  • Suitable for HPLC, GC and MS applications
Catalog No. 50-004-12882