Learn More
BURKLE INC Flow indicator CheMobil, PMP, blue
Supplier:  BURKLE INC 87202005
Encompass_Preferred
Flow indicators give precise visual indication of the flow velocity of liquids or gases being transported through hose connections. The flow indicator with impeller functions in both directions and in any position. The impeller (PP) starts to rotate at a flow rate about 150 ml/min (5.07 oz./min) (liquids).
Catalog No. 50-000-26099