Learn More
AVANTOR MASTERFLEX (VWR LLC) Dynalon Ball Flow Indicator, 0.2 to 6.5 L/min for 1/4 to 3/8"" ID Tubing
Supplier:  AVANTOR MASTERFLEX (VWR LLC) MFLX0629700
Encompass
Indicators give visual confirmation of flow through a clear polystyrene housing. All models can be used with noncorrosive liquids and gases. Connect tubing to indicators via hose barb connections; model 06297-05 has straight ends. All models are pressure rated to 10 psig.
Catalog No. NC9100614