Learn More
AVANTOR MASTERFLEX (VWR LLC) Masterflex Variable-Area In-Line Flowmeter, Direct Reading, Acrylic Housing, 1/4"" NPT(F); 0.25 GPM Water
Supplier:  AVANTOR MASTERFLEX (VWR LLC) MFLX3247700
Encompass
These meters are designed for use in more aggressive process environments. In addition to the machined acrylic bodies, the design utilizes stainless steel float guides. The combination is a meter suitable for high-vibration, high-fluctuation process applications. Some versions include polypropylene end fittings reinforced with aluminum stress rings for added strength to the meters¿ design.
Catalog No. NC1028837