Learn More
Anton Paar Flow module
Supplier:  Anton Paar 317785
Encompass_Preferred
Module is separate and removable- Unique technology allows especially fast and stabletemperature control- Facilitates the use of a dosing system, which can be used to addadditives to the sample and perform Litesizer DLS measurements inparallel and automatically
Catalog No. NC3893362
Add to Cart
Request a Quote