Learn More
Hach Company Flow-Thru CPVC Mt. Hard. Tee
Supplier:  Hach Company MH376
Encompass
Flow-Thru Mounting Hardware-PVC-pH/OrpPVC 1-1/2 Inch Tee and Aluminum Junction Box for Use with LCP pH/ORP Sensors
Catalog No. 50-002-07111