Learn More
Hach Company Flow-Thru CPVC Mt. Hard. Tee
Supplier:  Hach Company MH333N3NZ
Encompass
Flow-Thru CPVC Mounting Hardware Tee- 75 Each tee is a standard 3/4-inch tee with 3/4-inch NPT threads on all three openings
Catalog No. 50-002-07231