Learn More
AVANTOR MASTERFLEX (VWR LLC) Great Plains Industries A109GMN100NA1 Flowmeter/Totalizer, 3-50 GPM, 1"" NPT(F), Nylon
Supplier:  AVANTOR MASTERFLEX (VWR LLC) MFLX0561004
Encompass
A low-cost solution to light-duty process applications, this Flowmeter/Totalizer is battery-powered to eliminate the need for expensive wiring. It has two totalizers to allow for process or period-specific volume monitoring.
Catalog No. 50-631-046