Learn More
AVANTOR MASTERFLEX (VWR LLC) Masterflex Variable-Area Flowmeter, Correlated Reading, 316 SS Housing and Fittings, 150-mm Scale; 1682 mL/min Air
Supplier:  AVANTOR MASTERFLEX (VWR LLC) MFLX0326919
Encompass
Aluminum is economical and good for general use with noncorrosive gases and liquids. Brass is economical and good for use with water. Stainless steel withstands higher temperature and pressure and features excellent chemical compatibility. PTFE has the best chemical compatibility.
Catalog No. 50-196-1127