Learn More
AVANTOR MASTERFLEX (VWR LLC) Masterflex Variable-Area In-Line Dual Media/Scale Flowmeters with Valve, Direct-Reading, PTFE Fitting; 1.5 GPM Air/Water
Supplier:  AVANTOR MASTERFLEX (VWR LLC) MFLX3205322
Encompass
These meters have been designed to measure aggressive water-like liquid solutions and aggressive air-like gase mixtures. The dual water scale shows flow rate in gallons per minute (gpm) and liters per minute (LPM); the dual air scale shows flow rate in standard cubic feet per minute (scfm) and liters per minute (LPM). Flowmeters are individually tested to 1 x 10-7 sccs of helium or better.
Catalog No. 50-196-1185