Learn More
AVANTOR MASTERFLEX (VWR LLC) Masterflex Variable-Area Flowmeter, Direct Reading, Stainless Steel Fittings, In-line with Valve; 28 scfm Oxygen
Supplier:  AVANTOR MASTERFLEX (VWR LLC) MFLX3260519
Encompass
Designed for medium flow range applications, these flowmeters incorporate precision-machined glass and brass or stainless steel fittings to provide accurate and economical flow metering solutions. Simply rotate glass flowtube to read the gas you will be using¿oxygen, nitrogen, helium, carbon dioxide, and argon.
Catalog No. 50-196-1202