Learn More
Medchemexpress LLC Floxuridine | 50-91-9 | MFCD00006530 | 99.9% | 246.19 g/mol | C9H11FN2O5 | 10 MG
SDP
Supplier:  Medchemexpress LLC HYB0097R10MG
Encompass_Preferred
Floxuridine (5-fluorouracil 2'-deoxyriboside) analytical standard intended as a high-purity reference material for research and analytical applications. Supplied as a 10 mg vial in a brown glass bottle and stored sealed at 4°C to protect from light and moisture. CAS 50-91-9; molecular formula C9H11FN2O5; molecular weight 246.19 g/mol.
  • High purity (approximately 99.9%) suitable for analytical reference standards.
  • Intended for research and analytical applications such as assay calibration.
  • Supplied as a small-dose reference material (10 mg) in brown glass packaging.
  • Stable when stored sealed at 4°C, protected from light and moisture.
  • Useful for method development, calibration, and quantitative analysis.
Catalog No. 50-002-96821