Learn More
Wilmad Labglass FLSK RB 2N 1L 29/42C 24/40S
Supplier:  Wilmad Labglass LG7291256
Encompass_Preferred
Boiling Flask, Round Bottom, 2-Neck, 1000mL capacity. Joint Sizes: Center Neck: [ST] 29/42 outer; Angled Side Neck: [ST] 24/40 outer. Side neck is angled 20degree to the center neck.
Catalog No. 50-805-120