Learn More
Wilmad Labglass FLSK RB 3N 25ML 14/20C 14/20S
Supplier:  Wilmad Labglass ML1130700
Encompass_Preferred
Boiling Flask, Round Bottom, 3-Neck. Capacity: 25mL. Joint Sizes: Center Neck: [ST] 14/20; Side Necks: [ST] 14/20. Round bottom 3-neck boiling flask with vertical center neck and two side necks that are 180degree apart.
Catalog No. 50-807-413
Add to Cart
Description
Description
Product Suggestions
Product Suggestions
Videos
Safety and Handling
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov