Learn More
Wilmad Labglass FLSK RB 3N 500ML 24/40 24/40S
Supplier:  Wilmad Labglass LG7330186
Encompass_Preferred
Boiling Flask, Round Bottom, 3-Neck, 500mL capacity. Joint Sizes: Center Neck: [ST] 24/40; Vertical Side Necks: [ST] 24/40. Round bottom three neck boiling flask with standard taper joints on the center neck and the vertical side necks.
Catalog No. 50-804-370