Learn More
Wilmad Labglass FLSK RB 4N 12L 45/50C 24/40S
Supplier:  Wilmad Labglass LG7370212
Encompass_Preferred
Boiling Flask, Round Bottom, 4-Neck, 12 Liter capacity. Joint Sizes: Center Neck:[ST] 45/50; Vertical Side Necks: [ST] 24/40. Round bottom four neck boiling flask with center neck and three vertical side necks that are 90degree apart.
Catalog No. 50-805-325