Learn More
Wilmad Labglass FLSK RB 4N 5L 45/50C 29/42S
Supplier:  Wilmad Labglass LG7370270
Encompass_Preferred
Boiling Flask, Round Bottom, 4-Neck, 5000mL capacity. Joint Sizes: Center Neck:[ST] 45/50; Vertical Side Necks: [ST] 29/42. Round bottom four neck boiling flask with center neck and three vertical side necks that are 90degree apart.
Catalog No. 50-805-917
Item Discontinued This item has been discontinued by the Fisher Scientific channel. Please to view product availability in your area.