Learn More
Chemglass Life Sciences 500mL Shake Flask, No Baffles, Delong Neck, with Stainless Steel Closure, Approx OD x Height (mm): 100 x 200
SDP
Supplier:  Chemglass Life Sciences CLS203806
Encompass_Preferred
500mL Shake Flask, No Baffles, Delong Neck, with Stainless Steel Closure, Approx OD x Height (mm): 100 x 200. Non-baffled flasks are ideal for growing, mixing and storing both aerobic and anaerobic cultures. Graduated to approximate volumes. Delong style neck is made to fit stainless steel or plastic closures, while reducing the amount of splashing that would reach the closure. Supplied with a stainless steel closure.
Catalog No. 50-143-666