Learn More
Wilmad Labglass FLSK VOL CL A 1000ML PLS STPR
Supplier:  Wilmad Labglass LG8116120
Encompass_Preferred
Volumetric Flask, Class A, To Contain, 1000mL +/- 0.30mL capacity. Stopper Size: 22.Calibrated to contain at 20degreeC.Flask size complies with Federal Specifications NNN-F-289 for Type I, Class A(Unserialized flasks) and ASTM E-288.
Catalog No. 50-803-334
Item Discontinued This item has been discontinued by the Fisher Scientific channel. Please to view product availability in your area.
View Alternate Product